missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Smart-Tracker™ Wireless Datalogging Module

Ensuring that the cold chain has been preserved, whether your products are in storage or in transit, just makes sense. With Thermo Scientific™ Smart-Tracker™ it’s made simple.

Marca:  Thermo Scientific™ ST101-100

Detalles adicionales : Peso : 0.05600kg Transport : UN number : 3091 Chem class : 9

 Ver más versiones de este producto

Código de producto. 15498290

  • 286.00€ / cada uno


Consúltenos : 902 239 303 
Añadir a la cesta

Descripción

Descripción

  • Track temperature wirelessly and control modules using your smart phone with the Smart-Tracker application
  • Monitor storage facilities, freezers, refrigerators, heating equipment and packages in transit†
  • Share information via the Cloud anywhere in the world
  • Bluetooth wireless connection for use in many types of container and enclosures
  • Internal sensor: -20° to +60°C or -40° to +85°C
  • External PT100 sensor: -200° to +200°C
Especificaciones

Especificaciones

Smart-Tracker wireless datalogging module with external temperature PT100 probe (3.5m cable length)
ST101-100
-80°C
Lithium
0.44 lb. (0.2kg)
Wireless Datalogging Module with External Probe
-200° to +200°C
Module, external sensor, lithium battery, two colored sensor rings and two side magnetic mount
Bluetooth Certification: FCC, CE
Ultra-low freezers, LN2 cryo, ovens
0.24 x 0.63 x 1.02 in. (6 x 16 x 26mm) [Shipping]
Videos
Documentos


† Subject to network speed and availability