Learn More
Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin
Product Code. 30130375 Shop All Thermo Scientific Products
Change view
Click to view available options
Description:
Magnetic, SOLAμ Collection Plate, with Soluble Trypsin
Magnetic, with Soluble Trypsin
Magnetic, without Soluble Trypsin
Unit Size:
1 set
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
30130375 Magnetic, without Soluble Trypsin 1 set
30130372 Magnetic, SOLAμ Collection Plate, with Soluble Trypsin 1 set
30130371 Magnetic, with Soluble Trypsin 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30130375 Supplier Thermo Scientific™ Supplier No. 60118101
 Restricted Item
Fecha estimada de envoi: 27-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Perform simple and easy streptavidin immunocapture for protein sample cleanup prior to protein digestion or analysis.
Obtain fast, simple, highly reproducible, and sensitive immunocapture using the high loading capacity of the Thermo Scientific™ SMART Digest™ Bulk Fractionation Kits: Streptavidin. These kits were designed to remove the challenges often associated with the process of protein cleanup by providing a simple and easy capture workflow.
The kit contains beads with an attached streptavidin moiety enabling a strong streptavidin-biotin bond with the appropriate biotinylated molecule with specific affinity for the target biomolecule. The unwanted interferences are then removed leaving a clean sample containing the target molecule. Once completed subsequent digestion can be facilitated with SMART Digest enzymes if required.
TRUSTED_SUSTAINABILITY
Specifications
Description Magnetic, without Soluble Trypsin
Capture Streptavidin
Cleavage C-terminal side of lysine and arginine amino acids
Enzyme Trypsin
Format Bulk Magnetic
For Use With (Application) Immunocapture Sample Preparation
Product Line SMART Digest
Type Immunocapture
Includes Bulk Magnetic SMART Digest Resin