Learn More
Small Volume Sample Cups, Clarified Polypropylene, Cylindrical Bottom, 9.1 x 23.6 mm (1.2 mL), 2000/pack
Product Code. 30322156
Change view
Click to view available options
Diameter (Metric) Outside:
9.1 mm
Unit Size:
Each
Product Code. Diameter (Metric) Outside Unit Size
30322156 9.1 mm Each
1 options
This item is not returnable. View return policy
Product Code. 30322156 Supplier VHG Labs Supplier No. VHGFAASC4MP
 Restricted Item
Fecha estimada de envoi: 16-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outside 9.1 mm
Quantity 2000/Pk.
Capacity (Metric) 1.2 mL
Material Clarified Polypropylene
Height (Metric) 23.6 mm