missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16397-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
The protein encoded by this gene is a member of the SMAD family, which transduces signals from TGF-beta family members. The encoded protein is activated by bone morphogenetic proteins and interacts with SMAD4. Two transcript variants encoding different isoforms have been found for this gene.Especificaciones
SMAD9 | |
Polyclonal | |
Unconjugated | |
SMAD9 | |
MAD homolog 9, MADH6, MADH9, Mothers against DPP homolog 9, SMAD 9, SMAD family member 9, SMAD8A, SMAD8B, SMAD9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4093, 55994, 85435 | |
-20°C | |
Liquid |
Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15198, O54835, Q9JIW5 | |
SMAD9 | |
SMAD9 Fusion Protein Ag9490 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |