missing translation for 'onlineSavingsMsg'
Learn More

SMAD4/DPC4 Ab-1, Mouse Monoclonal Antibody,Lab Vision™ Epredia™

Provide accurate, reproducible results with the Epredia™ SMAD4/DPC4 Ab-1, Mouse Monoclonal Antibody.

Marca:  Epredia MS-865-B0

Detalles adicionales : Peso : 1.00000kg

 Ver más versiones de este producto

Código de producto. 12672477

  • 163.00€ / 100 microlitros

Envío estimado: 29-06-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

DPC4 (also named SMAD4; small mothers against decapentaplegic deleted in pancreatic carcinoma). The family of SMAD molecules comprises essential mediators of transforming growth factor β (TGF-β) signaling. To date, seven members of this family have been identified, each of which plays a specific and separate role in mediating TGF-β superfamily gene transcription. At least two different members, Smad2 and Smad4/DPC4, have been implicated in human cancer and appear to have tumor-suppressor functions whose inactivation may play an important role in the histogenesis of pancreatic and colorectal malignancy and possibly other human cancers as well.

Host Species: Mouse

Clone: SMD46; same as DCS-46

Isotype: IgG1

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Purified recombinant DPC4 protein

Molecular Weight: 60kDa

Positive Control: Ls174T, HT29, Raji, Jurkat cells

Cellular Localization: Nuclear

Recommended for:

Western Blotting

Immunofluorescence

Immunoprecipitation (Native and denatured)

Note:

USA: RUO; Int'l: RUO

Especificaciones

Especificaciones

SMAD4/DPC4 Ab-1
Monoclonal
200μg/mL
Concentrated
Purified recombinant DPC4 protein
100μL
Cancer and Tumor Biology
Primary
Human
Immunofluorescence, Immunoprecipitation, Western Blot
SMD46; same as DCS-46
Biotin
Murine
IgG1
RUO
Primary
Monoclonal
Videos
SDS
Documentos
Certificados