Learn More
Motic™ SLR Adapter, Projection Lens, B3
Ideal microscope camera adapter for the popular and advanced B3 Microscope. Motic™ SLR Adapter, Projection Lens, B3 allows you to document images using a microscope designed to accept ASC achromatic super contrast and Plan achromatic objectives.
Marca:  Motic™ SW011362
Detalles adicionales : Peso : 0.70000kg
 Ver más versiones de este producto
Código de producto. 12598415
  • 93.96€ / cada uno
Fecha de envío estimada
del proveedor 01-10-2019
Añadir a la cesta
Descripción
Descripción
  • Facilitates photomicography.
  • Suitable for B3 Series Microscopes.
  • B3 Microscopes are designed for advanced laboratory work and employ ASC achromatic super contrast and plan achromatic objectives.
Especificaciones
Especificaciones
Projection Lens
Camera (SLR) Adapter
B3 Series Microscopes
2.5X