missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC16A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20676-1-AP-150UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is specific to SLC16A2.
The protein encoded by this gene is one of two human homologs of Saccharomyces cerevisiae Rad23, a protein involved in nucleotide excision repair (NER). This protein was shown to interact with, and elevate the nucleotide excision activity of 3-methyladenine-DNA glycosylase (MPG), which suggested a role in DNA damage recognition in base excision repair. This protein contains an N-terminal ubiquitin-like domain, which was reported to interact with 26S proteasome, as well as with ubiquitin protein ligase E6AP, and thus suggests that this protein may be involved in the ubiquitin mediated proteolytic pathway in cells.Especificaciones
SLC16A2 | |
Polyclonal | |
Unconjugated | |
SLC16A2 | |
AHDS, DXS128, DXS128E, MCT 7, MCT 8, MCT7, MCT8, Monocarboxylate transporter 7, Monocarboxylate transporter 8, MRX22, SLC16A2, XPCT | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20502, 259248, 6567 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O70324, P36021, Q8K1P8 | |
SLC16A2 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |