missing translation for 'onlineSavingsMsg'
Learn More

SLC13A3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 26182-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16814705

  • 319.00€ / 150 microlitros

Envío estimado: 22-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Mammalian sodium-dicarboxylate cotransporters transport succinate and other Krebs cycle intermediates. They fall into 2 categories based on their substrate affinity: low affinity and high affinity. Both the low- and high-affinity transporters play an important role in the handling of citrate by the kidneys. The protein encoded by this gene represents the high-affinity form. Alternatively spliced transcript variants encoding different isoforms have been found for this gene, although the full-length nature of some of them have not been characterized yet.
Especificaciones

Especificaciones

SLC13A3
0.35 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8WWT9, Q91Y63
hNaDC3, NaDC 3, NADC3, SDCT2, SLC13A3
Rabbit
IgG
150 μL
-20°C
Primary
114644, 64849
Western Blot
Unconjugated
SLC13A3
Liquid
SLC13A3
SLC13A3 Fusion Protein Ag24183
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Videos
SDS
Documentos
Certificados