Learn More
Thermo Scientific™ Software SkanIt™
Utilice la herramienta definitiva para el control del lector de microplacas y la gestión de datos.
Marca:  Thermo Scientific™ N18600
Detalles adicionales : Peso : 1.00000kg
 Ver más versiones de este producto
Código de producto. 15823547
  • 3400.00€ / cada uno
Consúltenos : 902 239 303 
Añadir a la cesta
Descripción
Descripción
  • Multi-instrument software platform compatible with all Thermo Scientific microplate readers
  • Open license software for unlimited installation to multiple computers
  • Research Edition for life sciences; Drug Discovery Edition with features for compliance with FDA 21 CFR Part 11
  • User-interface languages available: English, French, German, Spanish, Portuguese, Italian, Russian, Japanese, and Chinese
  • Optional separate user accounts for controlling access
  • Robotic automation interface
  • Searchable SkanIt Cloud Library with ready-to-use protocols including calculations
  • Measurement sessions are easy to setup and navigate
  • Intuitive Virtual Pipetting Tool assists in defining the plate layout
  • Capable of endpoint, kinetic, spectral, bottom-reading multipoint, and kinetic-spectral measurements
  • Single- or multiple-wavelength measurements and even multi-technology measurements
  • Results data show in real-time during the measurement
  • Merge data from multiple measurements into one kinetic data set
  • Transfer data to an Excel file with a single click
  • Manually or automatically export data to .xlsx, .pdf, .xml, and .txt file formats
  • Automatically load measurement data into a predefined Excel template file
  • SkanIt Software Upgrade Package is also compatible with the discontinued Fluoroskan Ascent, Fluoroskan Ascent FL, and Luminoskan Ascent microplate readers.
    • Note: the upgrade package is compatible with Ascent instruments that are equipped with internal software version 2.6.
Especificaciones
Especificaciones
SkanIt Software upgrade package, drug discovery edition
Fluoroskan, Fluoroskan FL, Luminoskan, Fluoroskan Ascent, Fluoroskan Ascent FL, Luminoskan Ascent
Videos