missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SIX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11562-1-AP-20UL
Detalles adicionales : Peso : 0.00010kg
Descripción
This antibody is a rabbit polyclonal antibody raised against full length SIX2 of human origin.
This gene is a member of the vertebrate gene family which encode proteins homologous to the Drosophila ′sine oculis′ homeobox protein. SIX2 is a transcription factor which, like other members of this gene family, may be involved in limb or eye development.Especificaciones
SIX2 | |
Polyclonal | |
Unconjugated | |
SIX2 | |
Homeobox protein SIX2, Sine oculis homeobox homolog 2, SIX homeobox 2, SIX2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10736, 20472, 366542 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Immunofluorescence, Western Blot | |
0.52 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q62232, Q9NPC8 | |
Six2 | |
SIX2 Fusion Protein Ag2124 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |