missing translation for 'onlineSavingsMsg'
Learn More

SIT1 Antibody (SIT-01), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Bio-Techne NB500-486R

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18213794

  • 524.82€ / 0.10 ml
Fecha estimada de envío: 19-08-2024
para ver el stock

Descripción

Descripción

SIT1 Monoclonal specifically detects SIT1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Especificaciones

Especificaciones

SIT1
Monoclonal
DyLight 550
50 mM sodium borate with 0.05% sodium azide
SIT1
Bacterially produced recombinant intracellular fragment of human SIT.
Primary
The antibody SIT-01 reacts with SHP2-interacting transmembrane adaptor protein (SIT) expressed exclusively in lymphoid organs.
Store at 4°C in the dark.
Western Blot, Flow Cytometry
SIT-01
Western Blot, Flow Cytometry
gp30/40, MGC125908, MGC125909, RP11-331F9.5, SHP2 interacting transmembrane adaptor, SHP2-interacting transmembrane adapter protein, SHP2-interacting transmembrane adaptor protein, signaling threshold regulating transmembrane adaptor 1, signaling threshold-regulating transmembrane adapter 1, SITMGC125910, suppression inducing transmembrane adaptor 1, Suppression-inducing transmembrane adapter 1
Mouse
0.1 mL
27240
Human
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales