missing translation for 'onlineSavingsMsg'
Learn More

SIRT3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10099-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16816263

  • 319.00€ / 150 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

SIRT3 is a member of the sirtuin family of proteins, homologs to the yeast Sir2 protein. Members of the sirtuin family are characterized by a sirtuin core domain and grouped into four classes. The functions of human sirtuins have not yet been determined; however, yeast sirtuin proteins are known to regulate epigenetic gene silencing and suppress recombination of rDNA. Studies suggest that the human sirtuins may function as intracellular regulatory proteins with mono-ADP-ribosyltransferase activity. The SIRT3 is included in class I of the sirtuin family.
Especificaciones

Especificaciones

SIRT3
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8R104, Q9NTG7
hSIRT3, SIR2 like protein 3, SIR2L3, SIRT3
Rabbit
IgG
150 μL
-20°C
Primary
23410, 293615, 64384
Western Blot
Unconjugated
SIRT3
Liquid
Sirt3
SIRT3 Fusion Protein Ag0148
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados