missing translation for 'onlineSavingsMsg'
Learn More

TNF (TNFA) Recombinant Cynomolgus Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the soluble form of cynomolgus TNF-α (Val 77-Leu 233) fused to an initial Met at the N-terminus

Marca:  Invitrogen 90018CNAE50

Detalles adicionales : Peso : 0.10000kg

 Ver más versiones de este producto

Código de producto. 15319696

  • 271.00€ / 50 microgramos


Fecha de envío estimada
del proveedor 07-03-2019
Añadir a la cesta

Includes: One vial of lyophilized protein

Descripción

Descripción

Tumor Necrosis Factor (TNF), recombinant cynomolgus protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cytotoxicity assay using L-929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D; the ED50 is typically 20-80pg/mL
  • Formulation: Lyophilized in 50mM Tris, 50mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Especificación

Especificación

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
17.4kDa
50μg
TNF (Tumor Necrosis Factor)
Tumor necrosis factor
Assay Standard
E. coli
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
TNF-alpha
TNF
Cytokines & Receptors
Untagged
Videos

For Research Use Only. Not for use in diagnostic procedures.