missing translation for 'onlineSavingsMsg'
Learn More

SELE (E-SELECTIN) Recombinant Rat Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the rat SELE (P98105-1) extracellular domain (Met 1-Pro 494) fused to a polyhistidine tag at the C-terminus

Marca:  Invitrogen 80033R08H250

Detalles adicionales : Peso : 0.10000kg

Código de producto. 15309546

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Includes: Five vials of lyophilized protein

Descripción

Descripción

Selectin E (SELE), recombinant rat protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells; when 5 x 104 cells/well are added to rat SELE coated plates (2μg/mL with 100μL/well), approximately 20-35% will adhere for 1 hour incubation at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Especificaciones

Especificaciones

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
5 x 50μg
<1 EU/μg
Sele–selectin E
His Tag
Other Proteins
Other Proteins
53kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
SELE
SELE
Mammalian
Lyophilized
≥90% by SDS-PAGE
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only. Not for use in diagnostic procedures.