missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ RBP4 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding human RBP4 (NP_006735.2), including the signal sequence (Met 1-Leu 201), fused to a polyhistidine tag at the C-terminus

Marca:  Thermo Scientific™ 10354H08H50

Detalles adicionales : Peso : 0.01000kg

Código de producto. 13495826

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Retinol binding protein 4, plasma (RBP4), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing functional assays. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to bind all-trans retinoic acid; the binding of retinoic acid results in the quenching of tryptophan fluorescence in RBP4; recombinant human RBP4 can bind >1.0μM all-trans retinoic acid
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Especificaciones

Especificaciones

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
23kDa
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Retinol binding protein 4, plasma
His Tag
Other Proteins
Other Proteins
5950
50μg
Recombinant
RBP4
RBP6
Mammalian
Lyophilized
≥85% by SDS-PAGE
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only. Not for use in diagnostic procedures.