missing translation for 'onlineSavingsMsg'
Learn More

PDCD1LG2 Recombinant Mouse Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse PDCD1LG2 (Q9WUL5) (Met 1-Arg 219) fused to the Fc region of human IgG1 at the C-terminus

Marca:  Invitrogen 50804M02H250

Detalles adicionales : Peso : 0.10000kg

 Ver más versiones de este producto

Código de producto. 15319276

  • 562.00€ / 250 microgramos


Fecha de envío estimada
del proveedor 07-03-2019
Añadir a la cesta

Includes: Five vials of lyophilized protein

Descripción

Descripción

Programmed cell death 1 ligand 2 (PDCD1LG2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human PDCD1-His (Mfr. No. 10377-H08H) at 10μg/mL (100μL/well) can bind mouse PDCD1LG2-Fc; the EC50 of mouse PDCD1LG2-Fc is 0.95μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Especificación

Especificación

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
49.5kDa
5 x 50μg
Other Proteins
B7-DC, Btdc, F730015O22Rik, mgC124039, mgC124040, PD-L2B7-DC, Btdc, F730015O22Rik, PD-L2
PDCD1LG2
CD Proteins
IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Programmed cell death 1 ligand 2
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.