missing translation for 'onlineSavingsMsg'
Learn More

H1F0 Recombinant Human Protein, N-His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human H1F0 (P07305) (Met 1-Lys 194) fused to a polyhistidinetag at the N-terminus

244.00€ - 386.00€

Especificaciones

Pureza ≥90% by SDS-PAGE
Método de purificación PAGE
Concentración de endotoxinas <1 EU/μg
Formato Lyophilized
Alias del gen HSPD1, CPN60, GROEL, HLD4, HSP60, HSP65, HuCHA60, SPG13
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15237385
Ver documentos
Invitrogen
10920H07E25
5 x 5μg 244.00€
25 microgramos Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío
24-08-2020
Añadir a la cesta
 
15267385
Ver documentos
Invitrogen
10920H07E50
50μg 386.00€
50 microgramos Ver productos alternativos
Ahorre hasta 
»
Fecha estimada de envío
24-08-2020
Añadir a la cesta
 
Descripción

Descripción

H1 Histone Family, Member 0 (H1F0), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 600mM NaCl, 1mM DTT, pH 8.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Especificaciones

Especificaciones

≥90% by SDS-PAGE
<1 EU/μg
HSPD1, CPN60, GROEL, HLD4, HSP60, HSP65, HuCHA60, SPG13
PAGE
Lyophilized
3005

For Research Use Only. Not for use in diagnostic procedures.