missing translation for 'onlineSavingsMsg'
Learn More

EPHA2 Recombinant Mouse Protein (without Catalytic Activity Domain), His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mouse EPHA2 (NP_034269.2) extracellular domain (Met 1-Asn 535) was expressed, fused with a polyhistidine tag at the C-terminus

Marca:  Invitrogen 50586M08H50

Detalles adicionales : Peso : 0.10000kg

 Ver más versiones de este producto

Código de producto. 15319016

  • 197.00€ / 50 microgramos


Fecha de envío estimada
del proveedor 08-04-2019
Añadir a la cesta

Includes: One vial of protein in suspension buffer

Descripción

Descripción

Eph Receptor A2 (EPHA2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized mouse EphA2 at 2μg/mL (100μL/well) can bind mouse EphrinA1 with a linear range of 0.16-20ng/mL; 2. Immobilized mouse EphA2 at 2μg/mL (100μL/well) can bind human EphrinA1 with a linear range of 0.8-20ng/mL
  • Formulation: 20mM Tris, 150mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Especificación

Especificación

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
50μg
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Proteins
Videos

For Research Use Only. Not for use in diagnostic procedures.