missing translation for 'onlineSavingsMsg'
Learn More

DOT1L Recombinant Human Protein, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human DOT1L (NP_115871.1) N-terminal segment (Gly 2-Lys 416) fused to two additional amino acids (Gly & Pro) at the N-terminus

Marca:  Invitrogen 11552HNCE250

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15298645

  • 1202.00€ / 250 microgramos


Fecha de envío estimada
del proveedor 08-03-2019
Añadir a la cesta

Descripción

Descripción

DOT1-like histone H3 methyltransferase (S. cerevisiae) (DOT1L), recombinant human protein is supplied in frozen format. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: This recombinant protein has not been tested
  • Formulation: 20mM HEPES, 150mM NaCl, 1mM EDTA, 15% glycerol, pH 7.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Especificación

Especificación

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
84444
PAGE
5 x 50μg
Truncated/Recombinant
Dot1-Like, Histone H3 Methyltransferase (S. cerevisiae)
Assay Standard
E. coli
Frozen
47.6kDa
≥85% by SDS-PAGE
Store at -80°C. Thaw protein on ice before use and avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
DKFZp586P1823, DOT1, KIAA1814, KMT4
DOT1L
Methylation & Demethylation Proteins
Untagged
Videos

For Research Use Only. Not for use in diagnostic procedures.