missing translation for 'onlineSavingsMsg'
Learn More

CD96 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of mouse CD96 (Q3U0X8) (Met 1-Met 536) fused to a polyhistidine tag at the C-terminus

Marca:  Invitrogen 50758M08H50

Detalles adicionales : Peso : 0.10000kg

 Ver más versiones de este producto

Código de producto. 15319246

  • 269.00€ / 50 microgramos


Fecha de envío estimada
del proveedor 08-04-2019
Añadir a la cesta

Includes: One vial of lyophilized protein

Descripción

Descripción

CD96 antigen (CD96), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse CD96 at 10μg/mL (100μL/well) can bind mouse PVR with a linear range of 6.4-160ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Especificación

Especificación

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
58kDa
50μg
Other Proteins
1700109I12Rik, Tactile
CD97
CD Proteins
His Tag
Lyophilized
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
CD96 Antigen
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.