missing translation for 'onlineSavingsMsg'
Learn More

CD24 Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human CD24 (NP_037362.1) without the pro peptide (Met 1-Gly 59) fused to the Fc region of human IgG1 at the C-terminus

151.00€ - 243.00€

Especificaciones

Pureza ≥90% by SDS-PAGE
Método de purificación PAGE
Concentración de endotoxinas <1 EU/μg
Formato Lyophilized
Alias del gen CD24A, FLJ22950, FLJ43543, mgC75043
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15267595 Ver documentos Invitrogen
11030H02H25
5 x 5μg 151.00€
25 microgramos Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 05-11-2019
Añadir a la cesta
 
15297595 Ver documentos Invitrogen
11030H02H50
50μg 243.00€
50 microgramos Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 05-11-2019
Añadir a la cesta
 
Descripción

Descripción

Cluster of Differentiation 24 (CD24), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Especificaciones

Especificaciones

≥90% by SDS-PAGE
<1 EU/μg
CD24A, FLJ22950, FLJ43543, mgC75043
PAGE
Lyophilized
100133941

For Research Use Only. Not for use in diagnostic procedures.