missing translation for 'onlineSavingsMsg'
Learn More

BCL2L2 Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the human BCL-W (Q92843-1) (Met 1-Thr 172) fused to a polyhistidine tag at the C-terminus

272.00€

Especificaciones

Pureza ≥90% by SDS-PAGE
Método de purificación PAGE
Formato Frozen
Alias del gen BCL2L2, BCL-W, BCL2-L-2, BCLW, KIAA0271
ID gen (Entrez) 599
Ver más especificaciones

Productos 1
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15254575 Ver documentos Invitrogen
10059H08E50
50μg 272.00€
50 microgramos Ver productos alternativos
Ahorre hasta 
»
Fecha de envío estimada
del proveedor 07-02-2020
Añadir a la cesta
 
Descripción

Descripción

BCL2-Like 2 (BCL2L2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: 25mM HEPES, 100mM KCl, 10% Glycerol, pH 7.4
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Especificaciones

Especificaciones

≥90% by SDS-PAGE
Frozen
599
PAGE
BCL2L2, BCL-W, BCL2-L-2, BCLW, KIAA0271

For Research Use Only. Not for use in diagnostic procedures.