missing translation for 'onlineSavingsMsg'
Learn More

ARSA Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ser 506) of mouse ARSA precursor (NP_033843.2) fused to the polyhistidine tag at the C-terminus

Marca:  Invitrogen 50018M08H5

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15302556

Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas


Descripción

Descripción

Arylsulfatase A (ARSA), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave p-Nitrocatechol Sulfate (PNCS); the specific activity is >100pmoles/min./μg
  • Formulation: Lyophilized in 25mM Tris, 150mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Especificaciones

Especificaciones

Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories
5μg
<1 EU/μg
Arylsulfatase A
His Tag
Other Proteins
Other Proteins
53.5kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Arsa, AS-A, ASA, AW212749, As-2, As2, C230037L18Rik, TISP73
ARSA
Mammalian
Lyophilized
≥90% by SDS-PAGE
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only. Not for use in diagnostic procedures.