missing translation for 'onlineSavingsMsg'
Learn More

ACHE Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™

This recombinant protein was expressed from a DNA sequence encoding the mouse ACHE (NP_033729.1) (Met 1-Leu 614) fused to a polyhistidine tag at the C-terminus

Marca:  Invitrogen 50543M08H5

Detalles adicionales : Peso : 0.10000kg

 Ver más versiones de este producto

Código de producto. 15308946

  • 147.00€ / 5 microgramos


Fecha de envío estimada
del proveedor 02-07-2019
Añadir a la cesta

Includes: One vial of lyophilized protein

Descripción

Descripción

Acetylcholinesterase (ACHE), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave acetylthiocholine; the specific activity is >250nM/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Especificaciones

Especificaciones

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
66.2kDa
5μg
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Acetylcholinesterase
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Mouse
Other Proteins
mE1a, mE1b, mE1c, mE1c-long, mE1d, mE1d', mE1e
ACHE
Other Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.