missing translation for 'onlineSavingsMsg'
Learn More

Varilla de plata, 6,35 mm de diámetro, 99,9 % (base metálica), Thermo Scientific Chemicals

Catalog Number 11376578
Click to view available options
Cantidad:
2 cm
10 cm
50 cm
Este artículo no se puede devolver. Vea la política de devoluciones
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
Este artículo no se puede devolver. Vea la política de devoluciones
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Silver rod
Nombre de nota 6.35mm (0.25 in.) dia.
Notas del porcentaje del ensayo (base metálica)
Cantidad 2 cm
Porcentaje de pureza 99.9%
Olor Characteristic
Peso ≈3.32g/cm
Formulario Varilla, ≈3,32 g/cm

RUO - Para uso exclusivo en investigación

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'