Learn More
Varilla de plata, 12,5 mm de diámetro, Premion™, 99,9985 % (base metálica), Thermo Scientific Chemicals
Catalog Number 11301139
Click to view available options
Cantidad:
2 cm
10 cm
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
silver
[Ag]
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Silver rod
Nombre de nota 12.5mm (0.492 in.) dia.
Notas del porcentaje del ensayo (base metálica)
Cantidad 10 cm
Porcentaje de pureza 99.9985%
Grado Premion™
Olor Characteristic
Peso ≈12.87g/cm
Formulario Varilla, ≈12,87 g/cm