missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Óxido de plata(I), grado eléctrico, Thermo Scientific Chemicals
Click to view available options
Cantidad:
250 g
1 kg
Descripción
For battery plates, and pollution control filters. It is used in the preparation of other silver compounds, and silver-oxide batteries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Especificaciones
Nombre del producto químico o material | Silver (I) oxide |
Punto de fusión | 230°C (decomposition) |
Color | Marrón |
Forma física | Polvo de malla -325 |
Cantidad | 1 kg |
Número UN | UN1479 |
Índice Merck | 14,8521 |
Información de solubilidad | Soluble in dilute HNO3 and ammonia; Practically insoluble in water; Insoluble in alcohol |
Formula Weight (peso de la fórmula) | 231.74 |
Grado | Electrónico |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'