missing translation for 'onlineSavingsMsg'
Learn More

Cromato de plata(I), 99,9 % (base metálica), Thermo Scientific Chemicals

Catalog Number p-7059783
Click to view available options
Cantidad:
5 g
25 g
100 g
Este artículo no se puede devolver. Vea la política de devoluciones
7784-01-2
Ag2CrO4
331.73
MFCD00003402
OJKANDGLELGDHV-UHFFFAOYSA-N
silver chromate, unii-zj5092lwu9, silver chromate vi, silberchromat, silver i chromate, 2ag.cro4, chromic acid h2cro4 , disilver 1+ salt, chromic acid h2cro4 , silver 1+ salt 1:2, chromic acid, disilver i salt, disilver dioxido dioxo chromium
62666
diplata;dioxido(dioxo)cromo
[O-][Cr](=O)(=O)[O-].[Ag+].[Ag+]
Este artículo no se puede devolver. Vea la política de devoluciones
7784-01-2
Ag2CrO4
331.73
MFCD00003402
OJKANDGLELGDHV-UHFFFAOYSA-N
silver chromate, unii-zj5092lwu9, silver chromate vi, silberchromat, silver i chromate, 2ag.cro4, chromic acid h2cro4 , disilver 1+ salt, chromic acid h2cro4 , silver 1+ salt 1:2, chromic acid, disilver i salt, disilver dioxido dioxo chromium
62666
diplata;dioxido(dioxo)cromo
[O-][Cr](=O)(=O)[O-].[Ag+].[Ag+]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Silver (I) chromate
Color Rojo a marrón
Forma física Polvo
Cantidad 5 g
Notas del porcentaje del ensayo (base metálica)
Fórmula lineal Ag2CrO4
Número UN UN1479
Índice Merck 14,8510
Formula Weight (peso de la fórmula) 331.73
Porcentaje de pureza 99.9%
Olor Odorless
Sensibilidad Light sensitive
Densidad 5.63 g/mL
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'