missing translation for 'onlineSavingsMsg'
Learn More

Escamas de plata, 80 % < 20 micrómetros, 99,9 % (base metálica), Thermo Scientific Chemicals

Catalog Number p-7049543
Click to view available options
Cantidad:
10 g
50 g
250 g
Este artículo no se puede devolver. Vea la política de devoluciones
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
plata
[Ag]
Este artículo no se puede devolver. Vea la política de devoluciones
7440-22-4
Ag
107.87
MFCD00003397
BQCADISMDOOEFD-UHFFFAOYSA-N
argentum, metal, atom, colloidal, silver, colloidal, silver, elemental, algaedyn, amalgum, epinall, silber
23954
CHEBI:9141
plata
[Ag]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Silver flake
Notas del porcentaje del ensayo (base metálica)
Número UN UN3077
Cantidad 10 g
Porcentaje de pureza 99.9%
Olor Characteristic
Tamaño de partículas 80% <20 Micron
Peso Scott density: 18g/cm3 to 28g/in3
Formulario Escamas

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'