missing translation for 'onlineSavingsMsg'
Learn More

Carbonato de plata, 99,5 % (base metálica), Thermo Scientific Chemicals

Catalog Number p-4407804
Click to view available options
Cantidad:
25 g
100 g
534-16-7
CH2Ag2O3
277.76
MFCD00003403
WIKQEUJFZPCFNJ-UHFFFAOYSA-N
carbonic acid; silver, carbonic acid disilver
25203940
ácido carbónico;plata
C(=O)(O)O.[Ag].[Ag]
534-16-7
CH2Ag2O3
277.76
MFCD00003403
WIKQEUJFZPCFNJ-UHFFFAOYSA-N
carbonic acid; silver, carbonic acid disilver
25203940
ácido carbónico;plata
C(=O)(O)O.[Ag].[Ag]

Silver carbonate is used in the production of silver powder in microelectronics, conducting polymers, and in tooth surface treatment to prevent cavities. It is also used as an antimicrobial agent for infection resistant surgical fabric materials. It is used in organic synthesis, and in biological specimen staining.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Silver carbonate
Punto de fusión 218°C (decomposition)
Forma física Polvo
Cantidad 25 g
Intervalo de porcentaje de ensayo 99.5% (metals basis)
Información de solubilidad Insoluble in water,acetone,acetate and alcohol. Freely soluble in dilute HNO3 and ammonia.
Formula Weight (peso de la fórmula) 275.75
Sensibilidad Light sensitive
Densidad 6.08 g/mL

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'