missing translation for 'onlineSavingsMsg'
Learn More

Acetato de plata, anhidro, 99 %, Thermo Scientific Chemicals

Catalog Number p-7048852
Click to view available options
Cantidad:
25 g
100 g
563-63-3
C2H3AgO2
166.912
MFCD00012446
CQLFBEKRDQMJLZ-UHFFFAOYSA-M
silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
11246
plata;acetato
CC(=O)[O-].[Ag+]
563-63-3
C2H3AgO2
166.912
MFCD00012446
CQLFBEKRDQMJLZ-UHFFFAOYSA-M
silver acetate, silver i acetate, silver 1+ acetate, silver monoacetate, unii-19pps85f9h, acetic acid, silver 1+ salt, acetic acid silver salt, silver 1+ ion acetate, acetic acid, silver 1+ salt 1:1, silver acetate ag oac
11246
plata;acetato
CC(=O)[O-].[Ag+]

It is a reagent in the laboratory as a source of silver ions lacking an oxidizing anion. It is a reagent for direct ortho-arylation, and for conversion of organohalogen compounds into alcohols. Woodward cis-hydroxylation reaction employs silver acetate and iodine for selective conversin of alkenes into cis-diols. Silver acetate is the more preferred reagent for facile carbonylation of primary and secondary amines. It is also employed in the preparation of highly reflective, conductive silvered polymer films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Silver acetate
Punto de fusión (decomposition)
Nombre de nota Anhydrous
Color Blanco a gris
Forma física cristalino/a/s
Cantidad 25 g
Fórmula lineal AgOOCCH3
Número UN UN3077
Beilstein EM>10,
Índice Merck 14,8505
Información de solubilidad Soluble in dilute HNO3; Solubility in water increases with temperature.
Formula Weight (peso de la fórmula) 166.92
Porcentaje de pureza 99%
Olor Odorless
Sensibilidad Light sensitive
Densidad 3.26 g/mL
Mostrar más Mostrar menos

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'