Learn More
Thermo Scientific™ SilTite™ Ferrule Kits
Product Code. 10007084 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Thermo Scientific DSQ GC-MS Interface
Thermo Scientific ISQ GC-MS Interface
Thermo Scientific TRACE 1300/1310 GC SSL Injectors
Diameter (Metric) Inside:
0.1 to 0.25 mm
0.32 mm
0.53 mm
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Diameter (Metric) Inside Unit Size
10007084 Thermo Scientific DSQ GC-MS Interface 0.1 to 0.25 mm Pack of 10
10177904 Thermo Scientific ISQ GC-MS Interface 0.1 to 0.25 mm Pack of 10
10578405 Thermo Scientific ISQ GC-MS Interface 0.32 mm Pack of 10
10609965 Thermo Scientific ISQ GC-MS Interface 0.53 mm Pack of 10
10792897 Thermo Scientific TRACE 1300/1310 GC SSL Injectors 0.1 to 0.25 mm Pack of 10
10661196 Thermo Scientific TRACE 1300/1310 GC SSL Injectors 0.32 mm Pack of 10
10086154 Thermo Scientific TRACE 1300/1310 GC SSL Injectors 0.53 mm Pack of 10
10097294 Thermo Scientific DSQ GC-MS Interface 0.32 mm Pack of 10
10724556 Thermo Scientific DSQ GC-MS Interface 0.53 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10007084 Supplier Thermo Scientific™ Supplier No. 290MT229
Fecha estimada de envoi: 11-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits.
Eliminate leaks with Thermo Scientific™ SilTite Ferrule Kits. The ferrule forms a strong, permanent, airtight seal around the capillary column.
  • SilTite metal ferrules and nuts are made from the same material and have the same thermal expansion coefficient
  • Base of the ferrule is flat and forms a perfect seal with the MS interface
  • The ferrule's temperature tolerance is well above the limit of the injector, MS interface or GC oven
  • Unlike other ferrules, do not need re-tightening after installation
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule Kit
Description 290 MT229
For Use With (Equipment) Thermo Scientific DSQ GC-MS Interface
Material SilTite Metal
Includes 2 SilTite Nuts, 10 Ferrules
Diameter (Metric) Inside 0.1 to 0.25 mm
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.