missing translation for 'onlineSavingsMsg'
Learn More

Desecante de gel de sílice, indicación, ACS, Thermo Scientific Chemicals

Catalog Number p-4554790
Click to view available options
Cantidad:
100 g
500 g
Este artículo no se puede devolver. Vea la política de devoluciones
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:21615
7631-86-9
O2Si
60.08
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
O=[Si]=O
Este artículo no se puede devolver. Vea la política de devoluciones
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:21615
7631-86-9
O2Si
60.08
MFCD00011232 MFCD00217788 MFCD00163736 MFCD00148266 MFCD00603035 MFCD02100519 MFCD06202255 MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
O=[Si]=O

Used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits.

Silica gel desiccant is used to dry the air in industrial compressed air systems. It is also used as a preservation tool to control relative humidity in museum and library exhibitions and storage. Further, it is employed in diagnostic test strips, inhalation devices, syringes, drug test kits and hospital sanitation kits. In addition to this, it is used as a stationary phase in chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Nombre del producto químico o material Silica gel desiccant
Color Azul
Forma física Granular
Material Silica Gel
Cantidad 100 g
Olor Odorless
Nombre IUPAC dioxosilano
Grado ACS

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'