Learn More
Scotch-Brite™ Sienna Diamond Floor Pads
Product Code. 17148860
Change view
Click to view available options
Diameter (Metric):
406 mm
432 mm
505 mm
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
17148860 432 mm Pack of 5
17128860 406 mm Pack of 5
17138860 505 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17148860 Supplier Scotch-Brite™ Supplier No. 7100200177
  • ONLINE EXCLUSIVE PRICE
    401.00€
    245.77€ / paquete de 5
    Ahorro 155.23€
    38% Descuento
Fecha estimada de envoi: 05-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
For cleaning and conditioning worn or dull marble, terrazzo, slate, and polished concrete flooring. When floor appearance reaches a medium shine, the Scotch Brite™ Purple Diamond Floor Pad Plus can be used to further enhance the floor's appearance.
  • Simultaneously cleans and polishes flooring to a brilliant gloss finish without the use of chemicals
  • Ideal for worn or dull marble, terrazzo, vinyl, slate, and polished concrete floors
  • Easy to use with no expensive or harmful chemicals needed, only water
  • Recommended for use with low and standard speed machines
TRUSTED_SUSTAINABILITY
Specifications
Product Type Floor Pad
Color Brown
Diameter (Metric) 432 mm
For Use With (Application) Burnishing
For Use With (Equipment) Low and Standard Speed Machines
Style Round