missing translation for 'onlineSavingsMsg'
Learn More

SHMT2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 11099-1-AP-150UL

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16817953

  • 319.00€ / 150 microlitros

Envío estimado: 27-10-2021

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Mammalian serine hydroxymethyltransferase (SHMT) is a tetrameric, pyridoxal phosphate (PLP)-dependent enzyme that catalyzes the reversible interconversion of serine and tetrahydrofolate to glycine and methylenetetrahydrofolate in the cytoplasm (cSHMT, SHMT1) and mitochondria (mSHMT, SHMT2). cSHMT preferentially supplies one-carbon units for thymidylate biosynthesis, depletes methylenetetrahydrofolate pools for S-adenosylmethionine (SAM) synthesis by synthesizing serine, sequesters 5-methyltetrahydrofolate, and inhibits SAM synthesis.
Especificaciones

Especificaciones

SHMT2
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P34897, Q9CZN7
GLYA, Serine methylase, SHMT, SHMT2
Rabbit
IgG
150 μL
-20°C
Primary
108037, 299857, 6472
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SHMT2
Liquid
SHMT2
SHMT2 Fusion Protein Ag1584
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documentos
Certificados