Learn More
Epredia™ Shandon™ Stainless-Steel Brush with Plastic Handle, 7.25 in. (18.4cm)
Product Code. 15294013 Shop All Epredia Products
Change view
Click to view available options
Length (English):
7.25 in.
Unit Size:
1 set
Product Code. Length (English) Unit Size
15294013 7.25 in. 1 set
1 options
This item is not returnable. View return policy
Product Code. 15294013 Supplier Epredia™ Supplier No. 3120034
Fecha estimada de envoi: 20-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Guaranteed to be free from defects in material and workmanship, Shandon Stainless-Steel Brushes come three to a pack.
Size: 7.25 in. (18.4cm)
TRUSTED_SUSTAINABILITY
Specifications
Length (English) 7.25 in.