Learn More
Epredia™ Shandon™ Nickel-Plated Scalpel Blade Handle, Holds No. 20-25 Blades
Product Code. 12667776 Shop All Epredia Products
Change view
Click to view available options
Size:
# 4
Unit Size:
Each
Product Code. Size Unit Size
12667776 # 4 Each
1 options
This item is not returnable. View return policy
Product Code. 12667776 Supplier Epredia™ Supplier No. 53410
  • ONLINE EXCLUSIVE PRICE
    68.50€
    58.90€ / paquete de 1
    Ahorro 9.60€
    14% Descuento
View all promotions
Fecha estimada de envoi: 05-04-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Securely hold blades with the Epredia™ Shandon™ Nickel-Plated Scalpel Blade Handle, constructed of rustless, nickel-plated steel #4. 6.25in. (15.9cm) long.
  • Properly hardened, tempered and passivated
  • Electronically polished and cleaned for a high-quality finish
  • Quality patterns, dimensions and finish held to exacting standards, producing the finest instruments available
  • Defect-free in material and workmanship
TRUSTED_SUSTAINABILITY
Specifications
Blade Material Nickel-plated Steel
Size # 4
For Use With (Equipment) Holds # 20 to # 25 Blade