missing translation for 'onlineSavingsMsg'
Learn More

SGE™ MEPS™ BINs

Available in needle styles to suit LC and GC applications

Marca:  SGE™ 2900105

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15289223

Este producto se ha descatalogado y ya no está en venta. Consulte el producto para saber si se ofrecen alternativas o póngase en contacto con nuestro equipo de Atención al Cliente en el número 902 239 303.


Descripción

Descripción

The MEPS approach to sample preparation is suitable for reversed phases, normal phases, mixed mode and ion exchange chemistries.

  • For GC applications: needle is 23 gauge, 0.63mm OD, cone point style
  • For LC applications: needle is 22 gauge, 0.72mm OD, LC point style
  • Typical BIN life is conservatively about 40-100 samples but actual life is dependant on the sample matrix
  • The dimensions of the sorbent bed ensure that the sorbent performance remains identical to conventional SPE devices used for extraction of similar samples
  • Sample preparation for complex biological samples is readily adapted to MEPS and reduces the volume of sample and reagents required for extraction when compared with conventional SPE and other “micro extraction” procedures
  • Needles are packed in a sealed blister pack of 5 needles to ensure their integrity
  • Suitable for use with eVol™ automated syringes

Especificación

Especificación

Silica
23G
MEPS development kit
MEPS
HTA 300A plus, Thermo Scientific™, Varian 8400 and 100μL CTC analytics system
60Å
One each C18, C8, C2, Silica, C8+SCX
45μm
Videos