Learn More
Motic™ SFC-11C Head
Product Code. 15495372
Change view
Click to view available options
Head Style:
Binocular, 45° Inclined
Unit Size:
Each
Product Code. Head Style Unit Size
15495372 Binocular, 45° Inclined Each
1 options
This item is not returnable. View return policy
Product Code. 15495372 Supplier Motic™ Supplier No. PX11400001
  • ONLINE EXCLUSIVE PRICE
    257.00€
    247.00€ / paquete de 1
    Ahorro 10.00€
    3% Descuento
 Restricted Item
Fecha estimada de envoi: 25-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Replacement or modular head suitable for SFC-11C Stereo Microscope. Motic™ SFC-11C Head facilitates precise optical alignment and assures a three-dimensional, upright image suitable for a wide range of applications from dissecting to examining printed circuit boards.
  • The SFC-11 is the ideal stereo microscope for both educational and industrial applications.
  • Heavy-duty rack and pinion focusing block with slip clutch and tension adjustment.
  • Inclined at 45° for comfort and fatigue reduction.
  • Available in three turret magnification combinations [1X/2X, 1X/3X, and 2X/4X], thus providing magnification choices for various applications.
  • The standard working distance of 95mm is sufficient for majority of applications without the need for addition of auxiliary objectives.
  • With a variety of accessories available, the SFC-11 can adapt to most requirements without the need for factory or third-party modifications.
  • Available stands range from compact and simple incident stands to boom stands to print circuit board related stands as well as having numerous arm choices to complement the stands in way that satisfies all requirements.
  • Different contrast options permit complex observations with a simple stereo system.
Specifications
Magnification Power 2X, 4X
Objective Types Objective Turret Type C
For Use With (Equipment) SFC-11C Stereo Microscope
Product Type Microscope Head
Model SFC-11C
Head Style Binocular, 45° Inclined
Eyepiece WF 10X/20 mm