Learn More
SF3A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA551439
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: TEDSLMPEEE FLRRNKGPVS IKVQVPNMQD KTEWKLNGQV LVFTLPLTDQ VSVIKVKIHE ATGMPAGKQK LQYEGIFIKD SNSLAYYNMA NGAVIHLALK ESGGRKK Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
SF3A1 Polyclonal antibody specifically detects SF3A1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
SF3A1 | |
Polyclonal | |
Unconjugated | |
Sf3a1 | |
1200014H24Rik; 5930416L09Rik; AI159724; pre-mRNA processing 21; pre-mRNA splicing factor SF3a (120 kDa subunit); PRP21; PRPF21; SAP 114; SAP114; Sf3a1; sf3a1 protein; SF3A120; si:dz150f13.2; spliceosome-associated protein 114; splicing factor 3 subunit 1; splicing factor 3A subunit 1; splicing factor 3a, subunit 1; splicing factor 3a, subunit 1, 120kD; splicing factor 3a, subunit 1, 120kDa; Unknown (protein for MGC:65786); wu:fc38e01; wu:fc50a11; wu:fj37c05; zgc:65786 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10291, 305479, 67465 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15459, Q8K4Z5 | |
Sf3a1 | |
Recombinant protein corresponding to Human SF3A1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only