missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SERPINB6 (Human) Recombinant Protein

Human SERPINB6 full-length ORF ( AAH01394, 1 a.a. - 376 a.a.) recombinant protein with GST-tag at N-terminal.

Marca:  Abnova™ H00005269-P01.25ug

Detalles adicionales : Peso : 0.00010kg

 Ver más versiones de este producto

Código de producto. 16199061

  • 458.00€ / 25 microgramos


Fecha estimada de envío
21-10-2020
Añadir a la cesta

Descripción

Descripción

  • Sequence: MDVLAEANGTFALNLLKTLGKDNSKNVFFSPMSMSCALAMVYMGAKGNTAAQMAQILSFNKSGGGGDIHQGFQSLLTEVNKTGTQYLLRVANRLFGEKSCDFLSSFRDSCQKFYQAEMEELDFISAVEKSRKHINTWVAEKTEGKIAELLSPGSVDPLTRLVLVNAVYFRGNWDEQFDKENTEERLFKVSKNEEKPVQMMFKQSTFKKTYIGEIFTQILVLPYVGKELNMIIMLPDETTDLRTVEKELTYEKFVEWTRLDMMDEEEVEVSLPRFKLEESYDMESVLRNLGMTDAFELGKADFSGMSQTDLSLSKVVHKSFVEVNEEGTEAAAATAAIMMMRCARFVPRFCADHPFLFFIQHSKTNGILFCGRFSSP
Especificaciones

Especificaciones

AAH01394
serpin peptidase inhibitor, clade B (ovalbumin), member 6
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SERPINB6
GST
5269
Wheat germ expression system
25 μg
CAP, DKFZp686I04222, MGC111370, MSTP057, PI6, PTI, SPI3
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
Documentos