missing translation for 'onlineSavingsMsg'
Learn More

Serpin I1/Neuroserpin Antibody (002), Alexa Fluor™ 532, Novus Biologicals™

Rabbit Monoclonal Antibody

Marca:  Bio-Techne NBP2-90745AF532

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18216298

  • 607.98€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

Serpin I1/Neuroserpin Monoclonal specifically detects Serpin I1/Neuroserpin in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

Serpin I1/Neuroserpin
Monoclonal
Alexa Fluor 532
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Has cross-reactivity in ELISA with: Human SERPINI1 No cross-reactivity in ELISA with: Human SERPINA1 Human SERPINA3 Human SERPINA5 Human SERPINA12 Human SERPINB6 Human SERPINB9 Human SERPINC1 Human SERPIND1 Human SERPINF2 Human SERPING1 Human SERPINI2
Store at 4°C in the dark.
IgG
Immunohistochemistry, Immunohistochemistry (Paraffin)
2
Immunohistochemistry, Immunohistochemistry-Paraffin
DKFZp781N13156, neuroserpin, Peptidase inhibitor 12, PI-12, PI12Serpin I1, serine (or cysteine) proteinase inhibitor, clade I (neuroserpin), member 1, serpin peptidase inhibitor, clade I (neuroserpin), member 1
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Serpin I1/Neuroserpin (O35684; Met1-Leu410).
0.1 mL
Biologically Active Proteins
5274
Mouse
Purified
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales