missing translation for 'onlineSavingsMsg'
Learn More

SERBP1 Antibody (SERBP1/3496), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody

Marca:  Novus Biologicals NBP2-79844AF647

Detalles adicionales : Peso : 0.00970kg

Código de producto. 18214126

  • 578.82€ / 0.10 ml
Fecha estimada de envío: 14-08-2024
para ver el stock

Descripción

Descripción

SERBP1 Monoclonal specifically detects SERBP1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones

Especificaciones

SERBP1
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
SERBP1
Recombinant fragment of human SERBP1 protein (around aa3-139) (exact sequence is proprietary) (Uniprot: Q8NC51)
0.1 mL
Primary
Store at 4°C in the dark.
Immunohistochemistry, Immunohistochemistry (Paraffin)
SERBP1/3496
Immunohistochemistry, Immunohistochemistry-Paraffin
CGI-55, CHD3IP, chromodomain helicase DNA binding protein 3 interacting protein, DKFZP564M2423, HABP4L, PAI-1 mRNA binding protein, PAI1 RNA-binding protein 1, PAI-RBP1DKFZp564M2423, PAIRBP1FLJ90489, plasminogen activator inhibitor 1 RNA binding protein, plasminogen activator inhibitor 1 RNA-binding protein, SERPINE1 mRNA binding protein 1, SERPINE1 mRNA-binding protein 1
Mouse
Protein A or G purified
RUO
Human
IgG2c κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales