Learn More
Macherey-Nagel™ Sea Sand
Product Code. 10219891 Shop All Macherey-Nagel Products
Change view
Click to view available options
Type:
Sea Sand
Unit Size:
1000g
Product Code. Type Unit Size
10219891 Sea Sand 1000g
1 options
This item is not returnable. View return policy
Product Code. 10219891 Supplier Macherey-Nagel™ Supplier No. 727423
  • ONLINE EXCLUSIVE PRICE
    62.55€
    47.36€ / 1000 g
    Ahorro 15.19€
    24% Descuento
 Restricted Item
En stock
Add to basket
Add to basket
This item is not returnable. View return policy
Use to prepare columns for use in low-pressure flash chromatography. Macherey-Nagel™ Sea Sand is acid-washed and calcined, and is available in 1kg bottle.
When packing a column for use in low-pressure flash chromatography, users must ensure that the adsorbent cannot be flushed from the column and that the adsorbent does not become suspended in the eluent. Sea sand is placed above and below the silica to ensure a stable gel bed.
  • A pad of glass fiber wadding is placed in front of the tap and a layer of sea sand (about 0.5–1.0cm) is filled in above it
  • Then the dry adsorbent is packed into the column and is settled down until it forms a stable gel bed
  • The silica bed is covered with another layer of sea sand (about 0.5–1.0cm) to avoid suspension of the silica surface
Specifications
Material Sea Sand, acid-washed and calcined
Type Sea Sand
For Use With (Application) Low Pressure Flash Chromatography
For Use With (Equipment) Flash Chromatography Glass Columns