Learn More
SCRN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Thermo Scientific PA554797
Detalles adicionales : Peso : 0.02000kg
Descripción
Immunogen sequence: SVFKPFIFGV GVAQAPQVLS PTFGAQDPVR TLPRFQTQVD RRHTLYRGHQ AALGLMERDQ DRGQQLQQKQ QDLEQEGLEA TQGLLAGEWA PPLWEL Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 76%.
SCRN2 Polyclonal antibody specifically detects SCRN2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Especificaciones
SCRN2 | |
Polyclonal | |
Unconjugated | |
SCRN2 | |
AV001119; D11Moh48; SCRN2; secernin 2; secernin-2; Ses2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
90507 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96FV2 | |
SCRN2 | |
Recombinant protein corresponding to Human SCRN2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only