Learn More
Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone
Product Code. 10550042
Change view
Click to view available options
Packaging Quantity:
1
Unit Size:
Each
Product Code. Packaging Quantity Unit Size
10550042 1 Each
1 options
This item is not returnable. View return policy
Product Code. 10550042 Supplier Lenz Laborglasinstrumente™ Supplier No. 01312524
  • ONLINE EXCLUSIVE PRICE
    21.55€
    11.42€ / paquete de 1
    Ahorro 10.13€
    47% Descuento
Fecha estimada de envoi: 04-01-2027
Add to basket
Add to basket
This item is not returnable. View return policy
Manufactured from borosilicate glass. Lenz Laborglasinstrumente™ Screwthread Tubes with NS Cone are perfect for cell cultures in the laboratory.
  • Acc. to DIN 12257 and in addition to standard
  • With DIN thread (GL) and cone(NS)
  • Made of DURAN™ tubing
Specifications
Material Borosilicate Glass (3.3)
Packaging Quantity 1
Thread Size 25