Learn More
GPE Scientific™ Schlenk Flask, Cylindrical with Glass Tap and Cone
Product Code. 15844884
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15844884 250 mL Each
15834884 25 mL Each
15854884 50 mL Each
15814884 100 mL Each
15864884 500 mL Each
15824884 1000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15844884 Supplier GPE Scientific™ Supplier No. SCG/250
Fecha estimada de envoi: 03-02-2027
Add to basket
Add to basket
This item is not returnable. View return policy
GPE Scientific™ Schlenk Flask, Cylindrical with Glass Tap and Cone are specially designed for use with Schlenk lines and vacuum manifolds and are available in different capacities.
  • Cylindrical, Schlenk-style flask
  • Includes glass tap and cone
  • Available in different capacities
  • Available with different joint sizes
Specifications
Capacity (Metric) 250 mL
For Use With (Equipment) Schlenk lines and vacuum manifolds
Includes Glass tap and cone
Product Type Cylindrical Schlenk flask with tap and cone
Shape Cylindrical
Socket Joint Joint size: 24
Bottom Shape Cylindrical