Learn More
Kartell™ Schilling Burette
Automatic plastic Schilling burette (according to din 12.700 standards, Class B). With push button for drop-by-drop delivery. PE reservoir with stable PE-LD base.
Marca: Kartell™ 0198100
Detalles adicionales : Peso : 0.50000kg
Descripción
Automatic plastic Schilling burette (according to din 12.700 standards, Class B). With push button for drop-by-drop delivery. PE reservoir with stable PE-LD base.
Especificaciones
50 ml | |
Bureta Schilling | |
DIN 12700 |
PMP, Polietileno | |
Clase B |