missing translation for 'onlineSavingsMsg'
Learn More

SCGB1A1 Recombinant Protein (P02), Abnova

Brand:  Abnova H00007356-P02.10ug

Additional Details : Peso : 0.00010kg

 View more versions of this product

Product Code. 16101742

  • 341.70€ / 10 microgramos
Envío estimado: 24-01-2022

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications

Specifications

AAH04481
Solution
7356
SCGB1A1 (Human) Recombinant Protein (P02)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SCGB1A1
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
33.44
8
Glutathione Sepharose 4 Fast Flow
10 ug
EICPSFQRVIETLLMDTPSSYEAAMELFSPDQDMREAGAQLKKLVDTLPQKPRESIIKLMEKIAQSSLCN
CC10/CC16/CCSP/UGB
SCGB1A1
Wheat Germ (in vitro)
GST
Videos
SDS
Documents
Certificates
Special Offers

Special Offers