missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SC65 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA568253
Detalles adicionales : Peso : 0.01000kg
Descripción
Predicted to react with Mouse and Rat samples.
This nucleolar protein was first characterized because it was an autoantigen in cases on interstitial cystitis. The protein, with a predicted molecular weight of 50 kDa, appears to be localized in the particulate compartment of the interphase nucleolus, with a distribution distinct from that of nucleolar protein B23. During mitosis it is associated with chromosomes. [provided by RefSeq, Jul 2008]Especificaciones
SC65 | |
Polyclonal | |
Unconjugated | |
P3H4 | |
Synaptonemal complex protein SC65, Leprecan-like protein 4, Nucleolar autoantigen No55, LEPREL4, NOL55, SC65 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10609, 59101, 66180 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q92791, Q8K2B0, Q64375 | |
P3H4 | |
Synthesized peptide derived from human SC65. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |